Learn More
EPCAM Mouse anti-Human, Clone: EGP40/826, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human EPCAM.
Manufacturer:  Abnova Corporation MAB13334
Catalog No. 89-934-096
Add to cart
Description
Description
This gene encodes a carcinoma-associated antigen and is a member of a family that includes at least two type I membrane proteins. This antigen is expressed on most normal epithelial cells and gastrointestinal carcinomas and functions as a homotypic calcium-independent cell adhesion molecule. The antigen is being used as a target for immunotherapy treatment of human carcinomas. Mutations in this gene result in congenital tufting enteropathy. [provided by RefSeq]
Specifications
Specifications
EPCAM
EGP40/826
Mouse monoclonal antibody raised against synthetic peptide of human EPCAM.
In 10mM PBS.
17-1A/323/A3/CD326/CO-17A/CO17-1A/EGP/EGP-2/EGP34/EGP40/ESA/Ep-CAM/GA733-2/HEA125/KS1/4/KSA/M4S1/MH99/MIC18/MK-1/MOC31/TACST-1/TACSTD1/TROP1/hEGP-2
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ
Liquid
EPCAM
A synthetic peptide corresponding to amino acids 20-60 at N-terminus region of human EPCAM.
Protein A/G purification
RUO
Primary
4072