Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCAM Mouse anti-Human, Clone: EGP40/826, Abnova™

Catalog No. 89934095 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-095 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-095 Supplier Abnova Corporation Supplier No. MAB13333
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human EPCAM.

This gene encodes a carcinoma-associated antigen and is a member of a family that includes at least two type I membrane proteins. This antigen is expressed on most normal epithelial cells and gastrointestinal carcinomas and functions as a homotypic calcium-independent cell adhesion molecule. The antigen is being used as a target for immunotherapy treatment of human carcinomas. Mutations in this gene result in congenital tufting enteropathy. [provided by RefSeq]

Specifications

Antigen EPCAM
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone EGP40/826
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human EPCAM.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ
Formulation In 10mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Alias 17-1A/323/A3/CD326/CO-17A/CO17-1A/EGP/EGP-2/EGP34/EGP40/ESA/Ep-CAM/GA733-2/HEA125/KS1/4/KSA/M4S1/MH99/MIC18/MK-1/MOC31/TACST-1/TACSTD1/TROP1/hEGP-2
Gene Symbols EPCAM
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 20-60 at N-terminus region of human EPCAM.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4072
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.