Learn More
EPCAM Mouse anti-Human, PE, Clone: SPM134, Abnova™
Mouse monoclonal antibody raised against native human EPCAM.
Supplier: Abnova Corporation MAB13017
Description
This gene encodes a carcinoma-associated antigen and is a member of a family that includes at least two type I membrane proteins. This antigen is expressed on most normal epithelial cells and gastrointestinal carcinomas and functions as a homotypic calcium-independent cell adhesion molecule. The antigen is being used as a target for immunotherapy treatment of human carcinomas. Mutations in this gene result in congenital tufting enteropathy. [provided by RefSeq]
Specifications
| EPCAM | |
| Monoclonal | |
| PE | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| 17-1A/323/A3/CD326/CO-17A/CO17-1A/EGP/EGP-2/EGP34/EGP40/ESA/Ep-CAM/GA733-2/HEA125/KS1/4/KSA/M4S1/MH99/MIC18/MK-1/MOC31/TACST-1/TACSTD1/TROP1/hEGP-2 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4072 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence | |
| SPM134 | |
| Mouse monoclonal antibody raised against native human EPCAM. | |
| In PBS (0.05% BSA, 0.05% sodium azide). | |
| EPCAM | |
| Small cell lung carcinoma cells. | |
| 100 Reactions | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.