Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EpCAM Polyclonal Antibody, Cyanine5, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS4889RCY5

This item is not returnable.
View return policy
Description
EpCAM Polyclonal antibody specifically detects EpCAM in Human, Mouse samples. It is validated for Flow CytometrySpecifications
EpCAM | |
Polyclonal | |
Cyanine5 | |
EPCAM | |
adenocarcinoma-associated antigen; CD326; cell surface glycoprotein Trop-1; DIAR5; EGP; EGP-2; EGP314; EGP40; EPCAM; Ep-CAM; EpCAM1; epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; ESA; GA733-2; gp40; hEGP314; HNPCC8; human epithelial glycoprotein-2; KS 1/4 antigen; KS1/4; KSA; Ly74; lymphocyte antigen 74; M1S2; M4S1; major gastrointestinal tumor-associated protein GA733-2; mEGP314; membrane component, chromosome 4, surface marker (35kD glycoprotein); MIC18; MK-1; panepithelial glycoprotein 314; protein 289A; Protein D5.7A; Tacsd1; Tacstd1; TROP1; Trop-1 protein; Tumor-associated calcium signal transducer 1 | |
KLH conjugated synthetic peptide derived from human EpCAM | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P16422, Q99JW5 | |
Rabbit | |
Protein A | |
RUO | |
17075, 4072 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction