Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCAM Rabbit anti-Human, Unconjugated, Clone: EGP40/2041R, Abnova™

Catalog No. 89337951 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-951 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-951 Supplier Abnova Corporation Supplier No. RAB00526
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

EPCAM Monoclonal antibody specifically detects EPCAM in Human samples. It is validated for IHC-P.

Specifications

Antigen EPCAM
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EGP40/2041R
Conjugate Unconjugated
Description epithelial cell adhesion molecule
Gene EPCAM
Gene Alias 17-1A, 323/A3, CD326, CO-17A, CO17-1A, EGP, EGP-2, EGP34, EGP40, ESA, Ep-CAM, GA733-2, HEA125, KS1/4, KSA, M4S1, MH99, MIC18, MK-1, MOC31, TACST-1, TACSTD1, TROP1, hEGP-2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 100-224 at extracellular domain of human TACSTD1 fragment
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4072
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.