Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EpCAM/TROP1 Antibody (EGP40/1384), PE/Cy7 - (ECD), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254333PECY7
Description
EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Human samples. It is validated for Flow Cytometry.Specifications
EpCAM/TROP1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
EPCAM | |
Recombinant fragment from the extracellular domain of human EpCAM/TROP1 protein (around aa100-224) (exact sequence is proprietary) (Uniprot: P16422 ) | |
0.1 mL | |
Primary | |
EGP40 is a 40-43kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
EGP40/1384 | |
Flow Cytometry | |
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
Mouse | |
Protein A or G purified | |
Cancer, Cancer Stem Cells, Tumor Biomarkers | |
4072.0 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction