Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EpCAM/TROP1 Antibody (EGP40/1556R) - (ECD), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$304.50 - $560.50

Specifications

Antigen EpCAM/TROP1
Clone EGP40/1556R
Concentration 0.2 mg/mL
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25326620
SDP
View Documents
Novus Biologicals
NBP25326620UG
20 μg
Each for $304.50
Only null left
Add to Cart
 
NB253266100
SDP
View Documents
Novus Biologicals
NBP253266100UG
100 μg
Each for $560.50
Only null left
Add to Cart
 
NB253266200
SDP
View Documents
Novus Biologicals
NBP253266200UG
200 μg N/A N/A N/A
Description

Description

EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

EpCAM/TROP1
0.2 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cancer Stem Cells
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
4072
Recombinant human TACSTD1 protein
Primary
Store at 4C.
EGP40/1556R
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Monoclonal
Purified
RUO
Human, Mouse (Negative), Rat (Negative)
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
TACSTD1
IgG κ
Protein A purified
42 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.