Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EpCAM/TROP1 Antibody (rEGP40/7334), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | EpCAM/TROP1 |
---|---|
Clone | rEGP40/7334 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 μg/million cells, Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Human, Canine, Feline samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
EpCAM/TROP1 | |
0.2 mg/ml | |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cancer Stem Cells, Tumor Biomarkers | |
10mM PBS with 0.05% BSA | |
4072 | |
IgG1 κ | |
Protein A or G purified |
rEGP40/7334 | |
Flow Cytometry 1-2 μg/million cells, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Canine, Feline | |
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
Recombinant full-length human EpCAM/TROP1 protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title