Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPCR Antibody (101), DyLight 650

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290740C
Description
EPCR Monoclonal specifically detects EPCR in Mouse samples. It is validated for ELISA.Specifications
EPCR | |
Monoclonal | |
DyLight 650 | |
APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EPCR (Q64695; Met1-Ser214). | |
0.1 mL | |
Cardiovascular Biology | |
10544 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
101 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction