Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPCR Antibody (OTI12H5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271607H
Description
EPCR Monoclonal specifically detects EPCR in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EPCR | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PROCR | |
Recombinant protein fragment corresponding to amino acids 1-210 of human EPCR (NP_006395) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI12H5 | |
Western Blot, Immunohistochemistry | |
APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial | |
Mouse | |
Affinity Purified | |
Cardiovascular Biology | |
10544.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction