Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPDR1 Antibody (11), DyLight 405 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306190V
Description
EPDR1 Monoclonal antibody specifically detects EPDR1 in Human samples. It is validated for ELISASpecifications
EPDR1 | |
Monoclonal | |
DyLight 405 | |
EPDR, ependymin related protein 1 (zebrafish), MERP1mammalian ependymin related protein 1, MERP-1Upregulated in colorectal cancer gene 1 protein, UCC1mammalian ependymin-related protein 1 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EPDR1 (Q9UM22-1; Met1-Trp224). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
11 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
54749 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction