Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPDR1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986844
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6844 Supplier Bioss Supplier No. BS5837R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a type II transmembrane protein that is similar to two families of cell adhesion molecules, the protocadherins and ependymins. This protein may play a role in calcium-dependent cell adhesion. This protein is glycosylated, and the orthologous mouse protein is localized to the lysosome. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 8.

Specifications

Antigen EPDR1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EPDR1
Gene Accession No. Q9UM22
Gene Alias AU040950; AV006210; Epdm2; EPDR; EPDR1; Epdr2; ependymin 2; ependymin related 1; ependymin related protein 1; ependymin related protein 1 (zebrafish); ependymin related protein 2; mammalian ependymin related protein 1; Mammalian ependymin-related protein 1; MERP1; MERP-1; Merp2; MERP-2; UCC1; upregulated in colorectal cancer gene 1; Upregulated in colorectal cancer gene 1 protein
Gene Symbols EPDR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EPDR1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54749
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.