Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPG5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538061
Description
Do not aliquot.

Specifications
EPG5 | |
Polyclonal | |
Unconjugated | |
EPG5 | |
Ectopic P granules autophage protein 5 homolog antibody, HEEW1 antibody, hEPG5 antibody | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100502841, 364902, 57724 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9HCE0, Q80TA9 | |
EPG5 | |
A synthetic peptide derived from human EPG5 (121-240aa) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction