Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPGN Antibody (RM0228-2L47) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | EPGN |
---|---|
Clone | RM0228-2L47 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
EPGN Monoclonal specifically detects EPGN in Mouse samples. It is validated for Western Blot.Specifications
EPGN | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
epigen, epithelial mitogen homolog (mouse), PRO9904, UNQ3072/PRO9904 | |
EPGN | |
Primary |
RM0228-2L47 | |
Western Blot | |
Unconjugated | |
Rat | |
Protein Kinase | |
Q924X1.1 | |
255324 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title