Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eph receptor A3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7032R

Description
EPHA3 (EPA receptor A3), also known as ETK, HEK, ETK1, HEK4, TYRO4, is belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EPHA3 binds ephrin-A ligands.Specifications
Eph receptor A3 | |
Polyclonal | |
Unconjugated | |
EPHA3 | |
AW492086; Cek4; EK4; End3; Eph receptor A3; Epha3; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ETK; ETK1; H TYRO4; HEK; Hek4; Human embryo kinase; human embryo kinase 1; Mek4; Rek4; testicular tissue protein Li 64; TYRO4; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor REK4; Tyrosine-protein kinase TYRO4 | |
Rabbit | |
Protein A | |
RUO | |
2042 | |
-20°C | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P29320 | |
EPHA3 | |
KLH conjugated synthetic peptide derived from human Eph receptor A3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction