Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA1 Antibody (043), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290732AF350
Description
EphA1 Monoclonal specifically detects EphA1 in Mouse samples. It is validated for ELISA.Specifications
EphA1 | |
Monoclonal | |
Alexa Fluor 350 | |
EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA1 (Q60750; Met1-Glu548). | |
0.1 mL | |
Protein Kinase | |
2041 | |
Mouse | |
Purified |
ELISA | |
43 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Mouse EphA6 Mouse EPHA7 Mouse EPHB3 Mouse EPHB4 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction