Learn More
Promega EPHA1 Kinase Enzyme System, 1mg
Catalog No. PRV3562 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3562 1 mg
1 options
Catalog No. PRV3562 Supplier Promega Supplier No. V3562
Add to Cart
Add to Cart
Request a Quote
Recombinant mouse EPHA1 (amino acids 569-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EPHA1 is a member of the Eph family of receptor tyrosine kinases that have been implicated in mediating developmental events.
  • Easily Screen and Profile EPHA1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C