Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130291
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130291 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130291 Supplier Invitrogen™ Supplier No. PA130291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

EphA1 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA1 is expressed in some human cancer cell lines and has been implicated in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene EPHA1
Gene Accession No. P21709
Gene Alias 5730453L17Rik; AL033318; Embryonic stem cell kinase; EPH; EPH receptor A1; EPH tyrosine kinase; EPH tyrosine kinase 1; EPHA1; ephrin receptor EphA1; ephrin type-A receptor 1; EPHT; EPHT1; erythropoietin-producing hepatoma amplified sequence; erythropoietin-producing hepatoma receptor; Esk; hEpha1; mEpha1; MGC163163; oncogene EPH; soluble EPHA1 variant 1; soluble EPHA1 variant 2; tyrosine-protein kinase receptor EPH; Tyrosine-protein kinase receptor ESK
Gene Symbols EPHA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 913-943 amino acids from the C-terminal region of human EphA1.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2041
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.