Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphA10 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5145428
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145428 20 μL
FEPA5145427 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145428 Supplier Invitrogen™ Supplier No. PA5145428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPHA10 is a receptor for members of the ephrin-A family. It binds to EFNA3, EFNA4 and EFNA5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene EPHA10
Gene Accession No. Q5JZY3, Q8BYG9
Gene Alias A330090H18; EPH receptor A10; EPHA10; EphA10s protein; ephrin type-A receptor 10; RGD1562679
Gene Symbols EPHA10
Host Species Rabbit
Immunogen Recombinant Human EPHA10 protein, Val645-Leu900 (Accession #Q5JZY3).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230735, 284656, 298528
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.