Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA2 Antibody (1C1) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2526770.025MG
Description
EphA2 Monoclonal specifically detects EphA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Agonist Activity, Immunofluorescence, CyTOF-ready.Specifications
EphA2 | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
EPHA2 | |
Soluble human sEphA2, residues 183-510. | |
0.025 mg | |
Agonist Activity | |
1969 | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, CyTOF | |
1C1 | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Agonist Activity, Immunofluorescence, CyTOF-ready | |
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
The antibody binds to human EphA2 with a Kd of 0.89 nM. | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction