Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA2 Antibody (1C1), Janelia Fluor™ 646, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP281069JF646
Description
EphA2 Monoclonal specifically detects EphA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
EphA2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
EPHA2 | |
1C1 was isolated after screening of a non-immunized human Fab phage library. Selection was based on the ability to bind soluble human recombinant EphA2 (residues 183-510). The Fab was subsequently converted to full length human IgG1. | |
0.1 mL | |
Cancer, Protein Kinase | |
1969 | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
1C1 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
Human | |
Protein A purified | |
RUO | |
Primary | |
1C1 binds specifically to the amino-termianal LBD (ligand-binding domain) region of the EphA2 ectodomain (IgG1 KD ∽0.8 nM; Fab KD ∽140 nM) and not to other related human Eph receptors such as EphA1, EphA2, and EphA4, or any EphB receptor family members. | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction