Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA2 Antibody (233720), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB639X
Description
EphA2 Monoclonal antibody specifically detects EphA2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
EphA2 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
233720 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA2, Ala22-Ala535 (predicted), Accession # AAA82113 | |
0.5 mL | |
Cancer, Protein Kinase | |
1969 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction