Learn More
EphA2 Antibody (371805), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3035MFV500
Description
EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF, KnockDownSpecifications
| EphA2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Protein Kinase | |
| 1969 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, CyTOF, KnockDown | |
| 371805 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated | |
| ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
| Mouse myeloma cell line NS0-derived recombinant human EphA2, Gln25-Asn534, Accession # P29317 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |