Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA2 Antibody (M088), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323321
Description
EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Immunofluorescence, ImmunoprecipitationSpecifications
EphA2 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunofluorescence, Immunoprecipitation | |
M088 | |
Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:100 | |
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
Clone (M088) was generated from a recombinant protein that included the extracellular region of human EphA2 protein. [UniProt# P29317] | |
0.1 mL | |
Cancer, Protein Kinase | |
1969 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction