Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | EphA2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224489SS
![]() |
Novus Biologicals
NBP224489SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224489
![]() |
Novus Biologicals
NBP224489 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
EphA2 Polyclonal specifically detects EphA2 in Human, Bovine, Canine, Equine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EphA2 | |
Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Rabbit | |
Cancer, Protein Kinase | |
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
EPHA2 | |
IgG | |
Affinity Purified | |
100% homologous in human (isoforms CRA_a). |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Bovine, Canine, Equine, Monkey | |
P29317 | |
1969 | |
A portion of amino acids 450-500 of human EphA2 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title