Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514574
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514574 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514574 Supplier Invitrogen™ Supplier No. PA514574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with non-human primate based on sequence homology.

EPHA2 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA2 is overexpressed in many types of human cancers. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene EPHA2
Gene Accession No. P29317, Q03145
Gene Alias ARCC2; AW545284; CTPA; CTPP1; CTRCT6; Eck; EPH receptor A2; Epha2; ephrin receptor EphA2; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; I79_019726; Myk2; Sek2; Sek-2; soluble EPHA2 variant 1; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor SEK-2
Gene Symbols EPHA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 30-60 amino acids from the N-terminal region of human EphA2.
Purification Method Size-exclusion - Dialysis, Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13836, 1969
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.