Learn More
Invitrogen™ EphA2 Recombinant Rabbit Monoclonal Antibody (101), PE

Description
May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.
Specifications
Specifications
| Antigen | EphA2 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 101 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.5% BSA and 0.09% sodium azide |
| Gene | EPHA2 |
| Gene Accession No. | Q03145 |
| Gene Alias | ARCC2; AW545284; CTPA; CTPP1; CTRCT6; Eck; EPH receptor A2; Epha2; ephrin receptor EphA2; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; I79_019726; Myk2; Sek2; Sek-2; soluble EPHA2 variant 1; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor SEK-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.