Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EphA2 Recombinant Rabbit Monoclonal Antibody (BLR136H)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | EphA2 |
---|---|
Clone | BLR136H |
Concentration | 40 μg/mL |
Content And Storage | 4°C |
Applications | Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544459
|
Invitrogen™
MA544459 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544460
|
Invitrogen™
MA544460 |
20 μL |
Each for $173.10
|
|
|||||
Description
EphA2 Recombinant Monoclonal Antibody for Western Blot, IP
EPHA2 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA2 is overexpressed in many types of human cancers. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).Specifications
EphA2 | |
40 μg/mL | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
P29317 | |
1969 | |
Recombinant protein representing the extracellular domain of Human ephrin type-A receptor 2 (residues 25-534). | |
Antibody |
BLR136H | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
EPHA2 | |
ARCC2; AW545284; CTPA; CTPP1; CTRCT6; Eck; EPH receptor A2; Epha2; ephrin receptor EphA2; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; I79_019726; Myk2; Sek2; Sek-2; soluble EPHA2 variant 1; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor SEK-2 | |
EPHA2 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title