Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA4 Antibody (11C13) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | EphA4 |
---|---|
Clone | 11C13 |
Concentration | LYOPH |
Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018051
![]() |
Novus Biologicals
NB110936040.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB11093604
![]() |
Novus Biologicals
NB11093604 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
EphA4 Monoclonal specifically detects EphA4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EphA4 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
2043 | |
IgG1 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
11C13 | |
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Protein Kinase | |
EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, TYRO1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1 | |
EPHA4 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title