Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA4 Antibody (11C13), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11093604F
Description
EphA4 Monoclonal specifically detects EphA4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EphA4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
EPHA4 | |
Mouse EphA4 (extracellular domain) | |
0.1 mL | |
Primary | |
In this format, this does not cross-react with recombinant mouse EphA2, EphA6, EphA7, EphA8 and EphB2, EphB3, EphB4 or EphB6. | |
Store at 4C in the dark. | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
11C13 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, TYRO1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1 | |
Rat | |
Protein A or G purified | |
Protein Kinase | |
2043 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction