Learn More
Invitrogen™ EphA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532754
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%); Dog, Panda, Horse (94%); Bovine, Bat, Hamster, Elephant, Pig, Opossum, Turkey, Chicken (88%); Mouse, Rat, Rabbit, Platypus, Lizard, Xenopus (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.
Specifications
EphA4 | |
Polyclonal | |
Unconjugated | |
EPHA4 | |
2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2043 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P54764 | |
EPHA4 | |
Synthetic 16 amino acid peptide from internal region of human EPHA4. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.