Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphA5 Monoclonal Antibody (46CT61.6.4)
GREENER_CHOICE

Catalog No. PIMA511159
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511159 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511159 Supplier Invitrogen™ Supplier No. MA511159
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EphA5 Monoclonal Antibody for Western Blot, IHC (P)

EPHA5 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e. g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e. g. ABL, FAK, JAK, and SRC families).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 46CT61.6.4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene EPHA5
Gene Accession No. P54756
Gene Alias AI854630; ARCC2; AW125296; brain-specific kinase; BSK; CEK7; CEK-7; CTPA; CTPP1; ECK; eck-like sequence 1; EHK1; EHK-1; EK7; Ekh1; Els1; Els1.; epa5; EPH homology kinase 1; Eph receptor A5; EPHA5; EPH-like kinase 7; Ephrin A2; Ephrin A5; Ephrin type-A receptor 5; EphrinA2; EphrinA5; HEK7; receptor protein-tyrosine kinase HEK7; Rek7; TYRO4; tyrosine-protein kinase receptor EHK-1
Gene Symbols EPHA5
Host Species Mouse
Immunogen Purified His-tagged EPHA5 protein fragment.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2044
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.