Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHA5 Mouse anti-Human, Clone: 46CT61.6.4, Abnova™

Catalog No. 89934105 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-105 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-105 Supplier Abnova Corporation Supplier No. MAB12324
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human EPHA5.

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen EPHA5
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 46CT61.6.4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human EPHA5.
Dilution Immunohistochemistry (1:10-1:50) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias CEK7/EHK1/HEK7/TYRO4
Gene Symbols EPHA5
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human EPHA5.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2044
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.