Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589694 Supplier Invitrogen™ Supplier No. PA589694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRGSGPRGAG RRRPPSGGGD TPITPASLAG CYSAPRRAPL WTCLLLCAAL RTLLASPSNE VNLLDSRTVM GDLGWIAFPK NGWEEIGEVD ENYAPIHTYQ VCKVMEQNQN NWLLTSWISN EGASRIFIEL; Positive Samples: U-87MG, U-251MG, Mouse brain, Rat brain; Cellular Location: Cell membrane, Cell projection, Single-pass type I membrane protein, axon, dendrite.

EPHA5 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e. g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e. g. ABL, FAK, JAK, and SRC families).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EPHA5
Gene Accession No. P54756, P54757, Q60629
Gene Alias AI854630; ARCC2; AW125296; brain-specific kinase; BSK; CEK7; CEK-7; CTPA; CTPP1; ECK; eck-like sequence 1; EHK1; EHK-1; EK7; Ekh1; Els1; Els1.; epa5; EPH homology kinase 1; Eph receptor A5; EPHA5; EPH-like kinase 7; Ephrin A2; Ephrin A5; Ephrin type-A receptor 5; EphrinA2; EphrinA5; HEK7; receptor protein-tyrosine kinase HEK7; Rek7; TYRO4; tyrosine-protein kinase receptor EHK-1
Gene Symbols EPHA5
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13839, 2044, 79208
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.