Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA6 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298326MFV610
Description
EphA6 Polyclonal antibody specifically detects EphA6 in Mouse samples. It is validated for ELISASpecifications
EphA6 | |
Polyclonal | |
ELISA | |
DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, EHK2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, PRO57066 | |
Produced in rabbits immunized with purified, recombinant Mouse EphA6 (Accession#: NP_031964.2; Met 1-Gln 546) | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
285220 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction