Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHA6 Mouse anti-Human, Clone:3D5B10, Abnova™

Catalog No. 89334437 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-437 Supplier Abnova Corporation Supplier No. MAB17692

Mouse monoclonal antibody raised against recombinant human EPHA6.

Specifications

Antigen EPHA6
Applications Western Blot
Classification Monoclonal
Clone 3D5B10
Conjugate Unconjugated
Description EPH receptor A6
Dilution ELISA (1:10000)
Flow Cytometry
Immunocytochemistry
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene EPHA6
Gene Alias DKFZp434C1418, EPA6, FLJ35246, PRO57066
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 695-795 of human EPHA6 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 285220
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.