Learn More
Promega EPHA6-NanoLuc Fusion Vector, 20microg
Shop All Promega Corporation Products
Click to view available options
Quantity:
20 μg
Description
- Standard Cloning Vectors That Can Produce ssDNA and In vitro Transcribe Sense and Antisense RNA
- Blue/white screening for easy identification of recombinant clones
- In vitro transcription using the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
- The difference between + and − vectors is orientation of the f1 origin
Specifications
Specifications
| Quantity | 20 μg |
| Storage Requirements | -30°C to -10°C |