Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115187 Supplier Invitrogen™ Supplier No. PA5115187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EPHA6.

EPHA6 is a member of the ephrin receptor subfamily of protein-tyrosine kinases which have been implicated in axon guidance, neuron-target interactions, regional compartmentalization, and synaptic functions in nervous systems. EPHA6 is highly expressed in the brain and testis. Reduction in EPHA6 has been detected in Hypospadias, a common defect affecting the growth and closure of the external genitalia. Genetic inhibition of EPHA6 in mice produces behavioral deficits specifically in tests of learning and memory. Mice deficient in EPHA6 show reduced memory of the consequences of the training context.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Epha6
Gene Accession No. P54758, Q62413, Q9UF33
Gene Alias DKFZp434C1418; EHK2; EHK-2; EK12; EPA6; EPH homology kinase 2; Eph receptor A6; EPHA6; EPH-like kinase 12; ephrin receptor EphA6; ephrin type-A receptor 6; FLJ35246; HEK12; m-ehk2; PRO57066; tyrosine-protein kinase receptor EHK-2
Gene Symbols Epha6
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHA6(Accession Q9UF33), corresponding to amino acid residues S953-R1003.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13840, 285220, 29202
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.