Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA7 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298316MFV450
Description
EphA7 Polyclonal antibody specifically detects EphA7 in Mouse samples. It is validated for ELISASpecifications
EphA7 | |
Polyclonal | |
ELISA | |
EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
Produced in rabbits immunized with purified, recombinant Mouse EphA7 (Uniprot#: Q61772-1; Met 1-Ile 556) | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
2045 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction