Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Mouse EphA7 Antibody, R&D Systems™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
| Antigen | EphA7 | 
|---|---|
| Concentration | LYOPH | 
| Dilution | Western Blot 0.1 ug/mL | 
| Applications | Western Blot | 
| Classification | Polyclonal | 
Description
EphA7 Polyclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot.Specifications
| EphA7 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| Cek11, Ebk, EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, Mdk1, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
| EPHA7 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | 
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q61772 | |
| 2045 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA7 isoform 1 Ala30-Pro554 (Gly540-Glu544 del) Accession # Q61772 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse EphA7 in direct ELISAs and Western blots. In direct ELISAs, approximately 2% cross-reactivity with recombinant mouse (rm) EphA3 is observed and less than 1% cross-reactivity with rmEphA2, rmEphA4, rmEphA6, and rmEphA8 is observed. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            