Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPHA7 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A7E5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | EPHA7 |
---|---|
Clone | 1A7E5 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7084
![]() ![]() |
Proteintech Group Inc
666671IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.Specifications
EPHA7 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EHK 3, EHK3, EK11, EPH homology kinase 3, EPH like kinase 11, EPH receptor A7, EPHA7, Ephrin type A receptor 7, HEK11 | |
Epha7 | |
IgG1 | |
Protein G | |
Antibody |
1A7E5 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
P54759, Q15375, Q61772 | |
100154222, 13841, 171287, 2045 | |
EPHA7 Fusion Protein Ag3813 | |
Primary | |
-20°C | |
EPHA7 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title