Learn More
Invitrogen™ EphA7 Polyclonal Antibody

Description
PA1-28012 detects EphA7 from mouse samples.
Specifications
Specifications
| Antigen | EphA7 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and no preservative; pH 7.4 |
| Gene | EPHA7 |
| Gene Accession No. | Q61772 |
| Gene Alias | Cek11; Developmental kinase 1; Ebk; EHK3; EHK-3; EK11; embryonic brain kinase; epa7; EPH homology kinase 3; Eph homology kinase-3; EPH receptor A7; Epha7; EPHA7-S; EPH-like kinase 11; ephrin receptor EphA7; ephrin type-A receptor 7; HEK11; Mdk1; mDK-1; receptor protein-tyrosine kinase HEK11; tyrosine-protein kinase receptor EHK-3 |
| Gene Symbols | EPHA7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.