Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115188 Supplier Thermo Scientific Supplier No. PA5115188
Add to Cart
Add to Cart

EphA7 Polyclonal Antibody for Western Blot, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene EPHA7
Gene Accession No. P54759, Q15375, Q61772
Gene Alias Cek11; Developmental kinase 1; Ebk; EHK3; EHK-3; EK11; embryonic brain kinase; epa7; EPH homology kinase 3; Eph homology kinase-3; EPH receptor A7; Epha7; EPHA7-S; EPH-like kinase 11; ephrin receptor EphA7; ephrin type-A receptor 7; HEK11; Mdk1; mDK-1; receptor protein-tyrosine kinase HEK11; tyrosine-protein kinase receptor EHK-3
Gene Symbols EPHA7
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHA7(Accession Q15375), corresponding to amino acid residues C583-I633.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13841, 171287, 2045
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.