Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EphA8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556744
Description
Immunogen sequence: KRHCGYSKAF QDSDEEKMHY QNGQAPPPVF LPLHHPPGKL PEPQFYAQPH TYEEPGRAGR SFTREIEASR IHIEKIIGSG DSG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
Specifications
EphA8 | |
Polyclonal | |
Unconjugated | |
Epha8 | |
AW047546; Eek; EK3; epa8; EPH- and ELK-related kinase; EPH- and ELK-related tyrosine kinase; Eph receptor A8; Epha8; EPH-like kinase 3; Ephrin type-A receptor 8; Ephrin type-A receptor 8 precursor (Tyrosine-protein kinase receptor EEK) (EPH-and ELK-related kinase); Hek3; hydroxyaryl-protein kinase; KIAA1459; mKIAA1459; protein-tyrosine kinase; tyrosine-protein kinase receptor EEK; tyrosylprotein kinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2046 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P29322 | |
Epha8 | |
Recombinant protein corresponding to Human EphA8. Recombinant protein control fragment (Product #RP-94014). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction