Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB1 Antibody (022), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290032AF532
Description
EphB1 Monoclonal specifically detects EphB1 in Human samples. It is validated for ELISA.Specifications
EphB1 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human EPHB4 Human EPHB6 Mouse EphA1 Mouse EphA4 Mouse EphA6 Mouse EphA7 Mouse EphB3 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
22 | |
ELISA | |
EC 2.7.10, EC 2.7.10.1, EK6, ELK, EPH receptor B1, eph tyrosine kinase 2, EphB1, EPH-like kinase 6, ephrin type-B receptor 1, EPHT2FLJ37986, hEK6, NETHek6, soluble EPHB1 variant 1, Tyrosine-protein kinase receptor EPH-2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB1 (P54762-1; Met1-Pro540). | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase | |
2047 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction