Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB1/EphB3/EphB4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5106256
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106256 Supplier Thermo Scientific Supplier No. PA5106256

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EPH B1/3/4.

Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphB1/EphB3/EphB4
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHB4
Gene Accession No. P09759, P54753, P54754, P54760, P54761, P54762, Q8CBF3
Gene Alias 9330129L11; AI042935; AW456895; AW488255; C130099E04Rik; Cek10; Cek6; Developmental kinase 2; developmental kinase 5; EK2; EK6; Elk; Elkh; ELK-related protein tyrosine kinase; Embryonic kinase 2; ENSMUSG00000074119; EPH receptor B1; Eph receptor B2 (ELK-related protein tyrosine kinase); Eph receptor B3; Eph receptor B4; eph tyrosine kinase 2; Ephb1; Ephb2; EPHB3; Ephb4; EPH-like kinase 2; EPH-like kinase 6; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; ephrin receptor EphB4; ephrin type-B receptor 1; Ephrin type-B receptor 3; ephrin type-B receptor 4; EPHT2; Epth2; Erk; ETK2; HEK2; Hek6; Hepatoma transmembrane kinase; HTK; human embryo kinase 2; MDK2; mDK-2; Mdk5; mDK-5; Myk1; NET; Neuronally-expressed EPH-related tyrosine kinase; Sek4; soluble EPHB1 variant 1; soluble EPHB4 variant 1; soluble EPHB4 variant 2; soluble EPHB4 variant 3; Tyro11; TYRO6; Tyrosine kinase MYK-1; tyrosine-protein kinase receptor EPH-2; tyrosine-protein kinase receptor HTK; tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO11; tyrosine-protein kinase TYRO6
Gene Symbols EPHB1, EPHB3, EPHB4
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHB1(Accession P54762), corresponding to amino acid residues N755-S805.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13845, 13846, 2047, 2049, 2050, 24338, 270190, 287989, 686310
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.