Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    EphB2 Antibody (512012), DyLight 550, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB467L
Description
EphB2 Monoclonal antibody specifically detects EphB2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| EphB2 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified | 
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 512012 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CAPB, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, ERKHek5, HEK5, PCBC, protein-tyrosine kinase HEK5, Renal carcinoma antigen NY-REN-47, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphB2, Val27-Lys548 (predicted), Accession # P54763 | |
| 0.1 mL | |
| Prostate Cancer, Protein Kinase | |
| 2048 | |
| Store at 4°C in the dark. | |
| IgG2a | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            -DyLight-550-Product-Image-NBP2-34588R-img0001.jpg-250.jpg)