Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB2 Antibody (512012), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB467J
Description
EphB2 Monoclonal antibody specifically detects EphB2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
EphB2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
512012 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CAPB, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, ERKHek5, HEK5, PCBC, protein-tyrosine kinase HEK5, Renal carcinoma antigen NY-REN-47, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphB2, Val27-Lys548 (predicted), Accession # P54763 | |
0.1 mL | |
Prostate Cancer, Protein Kinase | |
2048 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction