Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB2 Mouse anti-Human, BB700, Clone: 2H9, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 746015
Description
EphB2 Monoclonal antibody specifically detects EphB2 in Human samples. It is validated for Flow Cytometry.
Specifications
EphB2 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
2H9 | |
Brilliant Blue 700 (BB700) | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction