Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB2 Mouse anti-Human, Brilliant Violet 605, Clone: 2H9, BD Optibuild™

Catalog No. BDB743764 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743764 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743764 Supplier BD Biosciences Supplier No. 743764
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2H9 monoclonal antibody specifically binds to the Ephrin Type-B Receptor 2 (EphB2). EphB2 is a type I transmembrane glycoprotein that belongs to the Eph receptor family of tyrosine kinase receptors. EphB2 serves as a cell surface receptor tyrosine kinase for membrane-anchored ligands referred to as type B ephrins (ephrin-B). Transmembrane ephrin-B family members are key regulators of embryogenesis including development of the nervous and vascular systems. The EphB2 receptor functions as a chemodirectant in cellular migration. EphB2/ephrin-B interactions orchestrate cell positioning by regulating cellular adhesion and repulsion during development, influencing cell fate, morphogenesis and organogenesis. In the adult body, Eph receptor signaling plays major roles in regulating the architecture and physiology of different tissues under normal as well as disease conditions. Ephrin-B1 and ephrin-B2 levels are upregulated in the vasculature during inflammation. Ephrin-B2 molecules that are localized to the luminal endothelial surface can signal through the EphB2 which is expressed by monocytes. This interaction promotes monocyte differentiation into proinflammatory macrophages. In the intestinal epithelium, EphB2/ephrin-B interactions regulate both cell positioning and tumor progression. These include intestinal stem cells (ISCs) which express high levels of EphB2. The 2H9 antibody reportedly blocks the interaction of EphB2 with ephrin ligands and crossreacts with mouse EphB2.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphB2
Classification Monoclonal
Clone 2H9
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias EPHB2, EphB2R, TYRO5, CAPB, DRT, EK5, EPHT3, EPTH3, HEK5, PCBC
Host Species Mouse
Immunogen Human EphB2 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.